Order Entry
Poland
ContactUsLinkComponent
ChromegaChiral(TM) Chiral CCO, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5530
Supplier:  Merck
ChromegaChiral(TM) Chiral CCO, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5530
Supplier:  Merck
Frequently Bought Together