ChromegaChiral(TM) Chiral CCA, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 25 cm x 30 mm 1 * 1 items
SIALES5427
: Merck
ChromegaChiral(TM) Chiral CCA, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 25 cm x 30 mm 1 * 1 items
SIALES5427
: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection