Antibodix(R) WCX-NP10 Weak Cation Exchange Column, L x I.D. 5 cm x 4.6 mm, 10 mum particle size, PEEK 1 * 1 items
SIALZ777275
: Merck
Antibodix(R) WCX-NP10 Weak Cation Exchange Column, L x I.D. 5 cm x 4.6 mm, 10 mum particle size, PEEK 1 * 1 items
SIALZ777275
: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection