Proteomix(R) WAX-NP10 Weak Anion Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 10 mum particle size 1 * 1 items
SIALZ777236
: Merck
Proteomix(R) WAX-NP10 Weak Anion Exchange Guard Cartridge and Holder, L x I.D. 1 cm x 4 mm, 10 mum particle size 1 * 1 items
SIALZ777236
: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection