Zenix(R) SEC-300 Gel Filtration Column, 3 mum, L x I.D. 30 cm x 7.8 mm, 3 mum particle size, stainless steel 1 * 1 items
SIALZ777033
: Merck
Zenix(R) SEC-300 Gel Filtration Column, 3 mum, L x I.D. 30 cm x 7.8 mm, 3 mum particle size, stainless steel 1 * 1 items
SIALZ777033
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK