Order Entry
Poland
ContactUsLinkComponent
Guiding Assembly P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK Rodaviss® with Ground Joint ns29/32 for shaft O. D. 8 mm, Ca t. No. C 540-08 and BOLA Magnetic Stirrer Heads P-MRK Rodaviss® with Ground Joint ns29/32 for shaft O. D. 10 mm, Cat. 1 * 1 items
Catalog # BOHLC921-05
Supplier:  Bohlender
undefined
Guiding Assembly P-MRK, Suitable for BOLA Magnetic Stirrer Heads P-MRK Rodaviss® with Ground Joint ns29/32 for shaft O. D. 8 mm, Ca t. No. C 540-08 and BOLA Magnetic Stirrer Heads P-MRK Rodaviss® with Ground Joint ns29/32 for shaft O. D. 10 mm, Cat. 1 * 1 items
Catalog # BOHLC921-05
Supplier:  Bohlender
Frequently Bought Together