Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Columns, HiTrap™ Capto PlasmidSelect

HiTrap™ Capto PlasmidSelect, 16×25 mm, 5 ml, <20 ml/min
Catalog # 17-5499-12
Supplier:   Cytiva
Columns, HiTrap™ Capto PlasmidSelect
Catalog # 17-5499-12
Supplier:  Cytiva
Columns, HiTrap™ Capto PlasmidSelect
Columns, HiTrap™ Capto PlasmidSelect

HiTrap™ Capto PlasmidSelect, 16×25 mm, 5 ml, <20 ml/min

Price
2 330 zł
Log in for availability
1 * 1 SZT
Product Details & Documents

Columns packed with Capto PlasmidSelect are suitable for purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications. Capto PlasmidSelect is a chromatography resin that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.

  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • Bioprocess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap columns

Capto PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production.

Specifications
  • Flow rate:
    <20 ml/min
  • Capacity:
    5 ml
  • Description:
    HiTrap™ Capto PlasmidSelect
  • Bed dimensions:
    16×25 mm
  • Cat. No.:
    17-5499-12
  • Max. pressure:
    5 bar