Microslide Superfrost 45° blue glass cello ISO 8037/1 1,0mm 1 * 50 items
Catalog # MENZAB00008132E01
Supplier: Epredia
Microslide Superfrost 45° blue glass cello ISO 8037/1 1,0mm 1 * 50 items
Catalog # MENZAB00008132E01
Supplier: Epredia
Frequently Bought Together
VWR®, Microscope Slides
Supplier: VWR Collection
Buckets with Snap-on Lids
Supplier: JOKEY PLASTIK
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Lens tissues
Supplier: GLASWARENFABRIK KARL HECHT
Sterican® Single-Use Needles for Special Applications
Supplier: B. Braun
Fine dosage syringes, Injekt®-F Solo
Supplier: B. Braun
Single use syringes, 2-piece, Injekt® Solo
Supplier: B. Braun
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cover Slips, Rectangular
Supplier: VWR Collection
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Trays for microscope slides
Supplier: LP ITALIANA
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF