Order Entry
Poland
Orders LinkContactUsLinkComponent
Polyester webbing sling, Protecta® DBI-SALA®, 3M™

Polyester webbing sling, Protecta® DBI-SALA®, 3M™

Polyester webbing sling with choke back, 45×900 mm
Catalog # CPSDKM419
Supplier:   CAPITAL SAFETY
Polyester webbing sling, Protecta® DBI-SALA®, 3M™
Catalog # CPSDKM419
Supplier:  CAPITAL SAFETY
Polyester webbing sling, Protecta® DBI-SALA®, 3M™
Polyester webbing sling, Protecta® DBI-SALA®, 3M™

Polyester webbing sling with choke back, 45×900 mm

Price
Inquire For Price
Log in for availability
1 * 1 SZT
Product Details & Documents

Sling polyester webbing and stainless steel links.

 : EN 795 Class-B

Specifications
  • Description:
    Polyester webbing sling with choke back, 45×900 mm
  • Seq:
    02
  • Cat. No.:
    CPSDKM419
  • Weight:
    0,47 kg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.