Order Entry
Poland
ContactUsLinkComponent
A LCN2 (Lipocalin2) ELISA with a senstivity of 0.188ng/ml and a range of 0.313-20ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
Catalog # GENOIT8307
Supplier:  G-Biosciences
A LCN2 (Lipocalin2) ELISA with a senstivity of 0.188ng/ml and a range of 0.313-20ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
Catalog # GENOIT8307
Supplier:  G-Biosciences
Frequently Bought Together