Order Entry
Poland
ContactUsLinkComponent
A Bnip3 (BCL2/adenovirus E1B 19 kDa protein-interacting protein 3) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8283
Supplier:  G-Biosciences
A Bnip3 (BCL2/adenovirus E1B 19 kDa protein-interacting protein 3) ELISA with a senstivity of 46.9pg/ml and a range of 78-5000pg/ml. Uses the detection method Sandwich ELISA, Double Antibody 1 * 1 KIT
Catalog # GENOIT8283
Supplier:  G-Biosciences
Frequently Bought Together