Order Entry
Poland
ContactUsLinkComponent
A CASP9 (Caspase 9) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
  GENOIT8026
A CASP9 (Caspase 9) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
  GENOIT8026
Frequently Bought Together