Order Entry
Poland
ContactUsLinkComponent
A CASP8 (Caspase 8) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
Catalog # GENOIT8025
Supplier:  G-Biosciences
A CASP8 (Caspase 8) ELISA with a senstivity of 0.094ng/ml and a range of 0.156-10ng/ml. Uses the detection method Sandwich ELISA, Do uble Antibody 1 * 1 KIT
Catalog # GENOIT8025
Supplier:  G-Biosciences
Frequently Bought Together