Order Entry
Poland
ContactUsLinkComponent
Propidium iodide (PI) is a red-fluorescent cell viability dye which is excluded from live cells as it is impermeable to the cell mem brane. The dye intercalates DNA and RNA of the dead cells to give red fluorescent signal as it can penetrate dead or 1 * 10 mL
Catalog # GENO786-1273
Supplier:  G-Biosciences
Propidium iodide (PI) is a red-fluorescent cell viability dye which is excluded from live cells as it is impermeable to the cell mem brane. The dye intercalates DNA and RNA of the dead cells to give red fluorescent signal as it can penetrate dead or 1 * 10 mL
Catalog # GENO786-1273
Supplier:  G-Biosciences
Frequently Bought Together