CONSUL MOUNT HISTOLOGY 1 * 1 ST
Catalog # SHAN9990440
Supplier: SHANDON
Frequently Bought Together
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
Supplier: Merck
pH indicator papers, universal/special
Supplier: MACHEREY-NAGEL
Sodium hydroxide solution 5 mol/L EMPROVE EXPERT 1 * 1 L
Supplier: MERCK PRODUCTION CHEMICALS
Test kits, AOX, Spectroquant®, Supelco®
Supplier: Merck
Reagents for copper determination using Lovibond® MD photometers
Supplier: Lovibond
RP500 MCART LAC 250 TEST 1 * 250 Tests
Supplier: BAYER VITAL
Colorimetric rapid tests, chlorine, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Powder pillows, silica
Supplier: Hach
Iron Standard Stock Solution
Supplier: CPACHEM
Sodium hydroxide 30% in water, Technical Grade
Supplier: Thermo Fisher Scientific
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach