KS135 Pa1/Alk1-Phenolphthalein Sub-Alk P 1 * 65 mL
Catalog # LOVI56L013565
Supplier: Lovibond
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Bottles, wide mouth, square without screw cap
Supplier: Kautex
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Measuring Beakers with Handle
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Hydrochloric acid 0.1 mol/l (0.1 N), AVS TITRINORM Reag. Ph. Eur. volumetric solution
Supplier: VWR Chemicals
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF