Methyl-4-hydroxybenzoate ≥98.0% NF
500 g
Catalog # SPCMME163-500GM
Supplier: Spectrum Chemical
Methyl-4-hydroxybenzoate ≥98.0% NF
Catalog # SPCMME163-500GM
Supplier: Spectrum Chemical
CAS Number: 99-76-3
Jump to:
Product Details & Documents
Methylparaben, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Exclamation mark.
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Storage Temperature:Ambient
Documents
Specifications
Specification Test Results
- Assay (C₈H₈O₃):98.0 - 102.0 %
- Color of Solution:To Pass Test
- Acidity:To Pass Test
- Residue On Ignition:max. 0.1 %
- Related Substances:To Pass Test
Specifications
- Pk:500 g



