CONOSTAN CUSTOM BLEND S-21+K+SB 500PPM 1 * 100 g
Catalog # PROOCON150-021-072
Supplier: LGC Standards PROMOCHEM
CONOSTAN CUSTOM BLEND S-21+K+SB 500PPM 1 * 100 g
Catalog # PROOCON150-021-072
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals