Order Entry
Poland
Orders LinkContactUsLinkComponent
1,8-Octanediol 98%

1,8-Octanediol 98%

Bottle, 100 g
Catalog # APOSOR2431-100G
Irritant
Supplier:   Apollo Scientific
1,8-Octanediol 98%
Catalog # APOSOR2431-100G
Supplier:  Apollo Scientific
CAS Number:  629-41-4
1,8-Octanediol 98%

Bottle, 100 g

CAS Number:  629-41-4
Price
319 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,8-Dihydroxy octane
  • CAS Number:629-41-4
  • Formula:C₈H₁₈O₂
  • MDL Number:MFCD00002989
  • Molecular Weight:146.23 g/mol
  • Boiling Pt:71…73 °C/20mmHg
  • Melting Pt:59…61 °C
  • Flash Pt:148 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.