Order Entry
Poland
ContactUsLinkComponent
Therm., similar Dr. Otte, similar DIN, +400+580:1°C, enclosed scale, cap. lens front, colourless, gallium filled, 370x7mm, Supremax- glass, Richter-top finish, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN59808-GA
Supplier:  Amarell
Therm., similar Dr. Otte, similar DIN, +400+580:1°C, enclosed scale, cap. lens front, colourless, gallium filled, 370x7mm, Supremax- glass, Richter-top finish, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARN59808-GA
Supplier:  Amarell
Frequently Bought Together