Glass therm., industrial, V-shape, DIN 16189, 0+200:2°C, built-in length 100mm, type B, special liquid filled, upper part 200mm, lar ge straight pattern, thread G 1/2 A 1 * 1 items
Catalog # AMART120092-FL
Supplier: Amarell
Frequently Bought Together