Order Entry
Poland
ContactUsLinkComponent
AMA Freezer therm., complete in plastic casing, -35+20:0,5C, c. 280x28mm, enclosed scale, cap. yellow backed, red alcohol filled, rounded top finish, suitable for government verification null
Catalog # AMART108406-FL
Supplier:  Amarell
AMA Freezer therm., complete in plastic casing, -35+20:0,5C, c. 280x28mm, enclosed scale, cap. yellow backed, red alcohol filled, rounded top finish, suitable for government verification null
Catalog # AMART108406-FL
Supplier:  Amarell
Frequently Bought Together