Order Entry
Poland
ContactUsLinkComponent
Therm. to set Amarell Universal, +297+360:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN71018-GA
Supplier:  Amarell
Therm. to set Amarell Universal, +297+360:0,1°C, 400x9mm, enclosed scale, cap.lens front, colourless, gallium filled, total immersio n, richter-top finish, suitable for government calibration 1 * 1 items
Catalog # AMARN71018-GA
Supplier:  Amarell
Frequently Bought Together