Order Entry
Poland
ContactUsLinkComponent
Thermometer similar to IP 74 C, solid stem, white backed, -35+70:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, 61mm immersion, 310mm long, null
Catalog # AMARA341050-CB
Supplier:  Amarell
Thermometer similar to IP 74 C, solid stem, white backed, -35+70:0,5C, capillary tube specially coated inside, with non-wetting bl ue special liquid, 61mm immersion, 310mm long, null
Catalog # AMARA341050-CB
Supplier:  Amarell
Frequently Bought Together