Order Entry
Poland
ContactUsLinkComponent
Distillation therm., similar to DIN 12779, 0+50:0,5°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersi on, length 210mm, pin-top finish with ring 1 * 1 items
  AMARD259600-FL
 :  Amarell
Distillation therm., similar to DIN 12779, 0+50:0,5°C, enclosed scale, cap. prismatic, colourless, red special liquid, total immersi on, length 210mm, pin-top finish with ring 1 * 1 items
  AMARD259600-FL
 :  Amarell
Frequently Bought Together