Thermometer similar to BS 593, B210C/100, 150+210:0,2°C, solid stem, white backed, gallium filled, 400mm long, 100mm immersion, dura ble pigment, with ice point scale, suitable for government verification 1 * 1 items
Catalog # AMARA403260-GA
Supplier: Amarell
Frequently Bought Together