Order Entry
Poland
ContactUsLinkComponent
Thermometer similar to BS 593, A130C/total, 100+130:0,1C, solid stem, white backed, capillary tube specially coated inside, with n on-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA403208-CB
Supplier:  Amarell
Thermometer similar to BS 593, A130C/total, 100+130:0,1C, solid stem, white backed, capillary tube specially coated inside, with n on-wetting blue special liquid, 400mm long, total immersion, durable pigment, with ice point scale, null
Catalog # AMARA403208-CB
Supplier:  Amarell
Frequently Bought Together