Tray, holding 100 x 2 mL vials, for fraction collector G1364E/F and G5664B 1 * 1 items
Catalog # HEWLG1364-44610
Supplier: Agilent
Tray, holding 100 x 2 mL vials, for fraction collector G1364E/F and G5664B 1 * 1 items
Catalog # HEWLG1364-44610
Supplier: Agilent
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Screw caps for certified screw top vials
Supplier: Agilent
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Inlet liners
Supplier: Agilent
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Headspace crimp caps with 20 mm septa
Supplier: Agilent
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals