Order Entry
Poland
ContactUsLinkComponent
Full tray, thermostattable for 4 well plates, for use with fraction collectors G1364B-F and G5664B 1 * 1 items
Catalog # HEWLG1364-60621
Supplier:  Agilent
undefined
Full tray, thermostattable for 4 well plates, for use with fraction collectors G1364B-F and G5664B 1 * 1 items
Catalog # HEWLG1364-60621
Supplier:  Agilent
Frequently Bought Together