Order Entry
Poland
ContactUsLinkComponent
General Purpose Pipettes Extra Small Bulb. Length 7,2 cm. Stem Diameter 3,3 mm. Overall Capacity 0,6 mL. Bulb Draw 0,3 mL 1 * 4.000 items
Catalog # SAMC281NL
Supplier:  SAMCO
undefined
General Purpose Pipettes Extra Small Bulb. Length 7,2 cm. Stem Diameter 3,3 mm. Overall Capacity 0,6 mL. Bulb Draw 0,3 mL 1 * 4.000 items
Catalog # SAMC281NL
Supplier:  SAMCO
Frequently Bought Together