Order Entry
Poland
Orders LinkContactUsLinkComponent
Human recombinant BAD

Human recombinant BAD

5 µG
Catalog # APOSBITP1468-5UG
Supplier:   Apollo Scientific
Human recombinant BAD
Catalog # APOSBITP1468-5UG
Supplier:  Apollo Scientific
CAS Number:  
Human recombinant BAD

5 µG

CAS Number:  
Price
1 940 zł
Log in for availability
1 * 5 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    5 µG
  • Cat. No.:
    APOSBITP1468-5UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    BAD
  • Species:
    Human

Specifications