Order Entry
Poland
Orders LinkContactUsLinkComponent
Rabbit Recombinant Prl

Rabbit Recombinant Prl

50 µG
Catalog # APOSBITP1249-50UG
Supplier:   Apollo Scientific
Rabbit Recombinant Prl
Catalog # APOSBITP1249-50UG
Supplier:  Apollo Scientific
CAS Number:  
Rabbit Recombinant Prl

50 µG

CAS Number:  
Price
983 zł
Log in for availability
1 * 50 µG
Product Details & Documents
Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    APOSBITP1249-50UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Prl
  • Protein synonyms:
    Prolactin
  • Species:
    Rabbit

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.