Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Rabbit Recombinant Prl

10 µG
  APOSBITP1249-10UG
Rabbit Recombinant Prl
  APOSBITP1249-10UG
 :  
Rabbit Recombinant Prl

10 µG

 :  
 
446 zł
   
1 * 10 µG
 
 

 

  • Pk:
    10 µG
  • Cat. No.:
    APOSBITP1249-10UG
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Prl
  • Protein synonyms:
    Prolactin
  • Species:
    Rabbit

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.