Order Entry
Poland
Orders LinkContactUsLinkComponent
 

3,3-Dimethoxyoxetane

Bottle, 1 g
Catalog # APOSOR312003-1G
Irritant
Supplier:   Apollo Scientific
3,3-Dimethoxyoxetane
Catalog # APOSOR312003-1G
Supplier:  Apollo Scientific
CAS Number:  922500-97-8
3,3-Dimethoxyoxetane

Bottle, 1 g

CAS Number:  922500-97-8
Price
1 800 zł
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:922500-97-8
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD12545828
  • Molecular Weight:118.13 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.