Order Entry
Poland
Orders LinkContactUsLinkComponent
2,4-Dimethoxy-5-fluoroaniline

2,4-Dimethoxy-5-fluoroaniline

Bottle, 1 g
Catalog # APOSPC7349-1G
Toxic
Supplier:   Apollo Scientific
2,4-Dimethoxy-5-fluoroaniline
Catalog # APOSPC7349-1G
Supplier:  Apollo Scientific
CAS Number:  195136-65-3
2,4-Dimethoxy-5-fluoroaniline

Bottle, 1 g

CAS Number:  195136-65-3
Price
495 zł
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  5-Fluoro-2,4-dimethoxyaniline
  • CAS Number:195136-65-3
  • Formula:C₈H₁₀FNO₂
  • MDL Number:MFCD12547861
  • Molecular Weight:171.17 g/mol
  • Melting Pt:47…49 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.