Order Entry
Poland
Orders LinkContactUsLinkComponent
4'-Nitroacetophenone 98%

4'-Nitroacetophenone 98%

Bottle, 5 kg
Catalog # APOSOR28938-5KG
Irritant
Supplier:   Apollo Scientific
4'-Nitroacetophenone 98%
Catalog # APOSOR28938-5KG
Supplier:  Apollo Scientific
CAS Number:  100-19-6
4'-Nitroacetophenone 98%

Bottle, 5 kg

CAS Number:  100-19-6
Price
1 330 zł
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  • CAS Number:100-19-6
  • Formula:C₈H₇NO₃
  • MDL Number:MFCD00007355
  • Molecular Weight:165.15 g/mol
  • Boiling Pt:201…202 °C
  • Melting Pt:75…78 °C
  • Flash Pt:149 °C
  • Storage Temperature:Ambient
  • EINECS:202-827-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.