Order Entry
Poland
Orders LinkContactUsLinkComponent
 

4'-Nitroacetophenone 98%

Bottle, 250 g
Catalog # APOSOR28938-250G
Irritant
Supplier:   Apollo Scientific
4'-Nitroacetophenone 98%
Catalog # APOSOR28938-250G
Supplier:  Apollo Scientific
CAS Number:  100-19-6
4'-Nitroacetophenone 98%

Bottle, 250 g

CAS Number:  100-19-6
Price
114 zł
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
  • CAS Number:100-19-6
  • Formula:C₈H₇NO₃
  • MDL Number:MFCD00007355
  • Molecular Weight:165.15 g/mol
  • Boiling Pt:201…202 °C
  • Melting Pt:75…78 °C
  • Flash Pt:149 °C
  • Storage Temperature:Ambient
  • EINECS:202-827-4
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g

Specifications