Order Entry
Poland
Orders LinkContactUsLinkComponent
 

3,3'-Dimethylbiphenyl

Bottle, 5 g
Catalog # APOSOR70107-5G
Irritant
Supplier:   Apollo Scientific
3,3'-Dimethylbiphenyl
Catalog # APOSOR70107-5G
Supplier:  Apollo Scientific
CAS Number:  612-75-9
3,3'-Dimethylbiphenyl

Bottle, 5 g

CAS Number:  612-75-9
Price
573 zł
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3,3'-Bitolyl, 3,3'-Dimethyl-1,1'-biphenyl
  • CAS Number:612-75-9
  • Formula:C₁₄H₁₄
  • MDL Number:MFCD00008534
  • Molecular Weight:182.27 g/mol
  • Boiling Pt:286 °C/713mmHg
  • Melting Pt:44382 °C
  • Density:0.999 g/cm³ (25 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications