Order Entry
Poland
Orders LinkContactUsLinkComponent
Silver nitrate 99.9+%

Silver nitrate 99.9+%

Bottle, 25 g
Catalog # APOSIN3235-25G
Oxidizing
Corrosive
Supplier:   Apollo Scientific
Silver nitrate 99.9+%
Catalog # APOSIN3235-25G
Supplier:  Apollo Scientific
CAS Number:  7761-88-8
Silver nitrate 99.9+%

Bottle, 25 g

CAS Number:  7761-88-8
Price
425 zł
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Melting Pt:212 °C (dec.)
  • Density:4.352 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.