Order Entry
Poland
Orders LinkContactUsLinkComponent
 

Benzaldehyde 99+%

Bottle
Catalog # APOSOR45085-100G
Irritant
Supplier:   Apollo Scientific
Benzaldehyde 99+%
Catalog # APOSOR45085-100G
Supplier:  Apollo Scientific
CAS Number:  100-52-7
Benzaldehyde 99+%

Bottle

CAS Number:  100-52-7
Price
213 zł
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

  • CAS Number:100-52-7
  • Formula:C₇H₆O
  • MDL Number:MFCD00003299
  • Molecular Weight:106,12194 g/mol
  • EINECS:202-860-4
  • UN:1990
Specifications

Specifications

  • Pack type:
    Bottle

Specifications