Nivalenol standard solution
Nivalenol, 100 µg/ml in acetonitrile, Bottle, 5 mL
Catalog # APOSBIMS126-5ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:5 mL
- Description:Nivalenol
- Cat. No.:APOSBIMS126-5ML
- Formula:C₁₅H₂₀O₇
- Description:Nivalenol
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:23282-20-4



