Fusarenon X Standard Solution
Fusarenon X, 100 µg/ml in acetonitrile, Bottle, 5 mL
Catalog # APOSBIMS116-5ML
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
Specifications
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:5 mL
- Description:Fusarenon X
- Cat. No.:APOSBIMS116-5ML
- Formula:C₁₇H₂₂O₈
- Description:Fusarenon X
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:23255-69-8
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



