Fusarenon X Standard Solution
Fusarenon X, 100 µg/ml in acetonitrile, Bottle, 1 mL
APOSBIMS116-1ML
Analytical reference standard solution in acetonitrile.
Caution: Expires in 12 months.
- Concentration:100 µg/ml in acetonitrile
- Pack type:Bottle
- Pk:1 mL
- Description:Fusarenon X
- Cat. No.:APOSBIMS116-1ML
- Formula:C₁₇H₂₂O₈
- Description:Fusarenon X
- Storage temperature:Freezer
- Flash point:2 °C
- CAS No.:23255-69-8
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



