Order Entry
Poland
Orders LinkContactUsLinkComponent
Ochratoxin A standard solution

Ochratoxin A standard solution

Ochratoxin A, 10 µg/ml in acetonitrile, Bottle, 5 mL
Catalog # APOSBIMS107-5ML
Supplier:   Apollo Scientific
Ochratoxin A standard solution
Catalog # APOSBIMS107-5ML
Supplier:  Apollo Scientific
CAS Number:  
Ochratoxin A standard solution

Ochratoxin A, 10 µg/ml in acetonitrile, Bottle, 5 mL

CAS Number:  
Price
4 210 zł
Log in for availability
1 * 5 mL
Product Details & Documents

Analytical reference standard solution in acetonitrile.

Caution: Expires in 12 months.

Documents

Specifications

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pack type:
    Bottle
  • Pk:
    5 mL
  • Description:
    Ochratoxin A

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.