Midostaurin
Bottle, 5 mg
APOSBIP1007-5MG
Semisynthetic derivate of Staurosporine. Inhibitor of PKC and cAMPK.
: PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide
- :120685-11-2
- :C₃₅H₃₀N₄O₄
- :MFCD12828879
- :570.64 g/mol
- :235…260 °C
- :Freezer
- Pack type:Bottle
- Pk:5 mg