Midostaurin
Bottle, 10 mg
Catalog # APOSBIP1007-10MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Semisynthetic derivate of Staurosporine. Inhibitor of PKC and cAMPK.
Synonyms: PKC412, N-((5S,6R,7R,9R)-6-Methoxy-5-methyl-14-oxo-6,7,8,9,15,16-hexahydro-5H,14H-17-oxa-4b,9a,15-triaza-5,9-methanodibenzo[b,h]cyclonona[jkl]cyclopenta[e]-as-indacen-7-yl)-N-methylbenzamide
- CAS Number:120685-11-2
- Formula:C₃₅H₃₀N₄O₄
- MDL Number:MFCD12828879
- Molecular Weight:570.64 g/mol
- Melting Pt:235…260 °C
- Storage Temperature:Freezer
Specifications
Specifications
- Pack type:Bottle
- Pk:10 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



