Order Entry
Poland
Orders LinkContactUsLinkComponent
Bacitracin

Bacitracin

Bottle, 5 g
Catalog # APOSBIB0106-5G
Irritant
Supplier:   Apollo Scientific
Bacitracin
Catalog # APOSBIB0106-5G
Supplier:  Apollo Scientific
CAS Number:  1405-87-4
Bacitracin

Bottle, 5 g

CAS Number:  1405-87-4
Price
142 zł
Log in for availability
1 * 5 g
Product Details & Documents

Bacitracin is active against gram-positive bacteria.

  • Approx. 75,000 units/g
  • CAS Number:1405-87-4
  • Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL Number:MFCD00062640
  • Molecular Weight:1421.60 g/mol
  • Melting Pt:221…225 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications