Order Entry
Poland
Orders LinkContactUsLinkComponent
Bacitracin

Bacitracin

Bottle, 25 g
Catalog # APOSBIB0106-25G
Irritant
Supplier:   Apollo Scientific
Bacitracin
Catalog # APOSBIB0106-25G
Supplier:  Apollo Scientific
CAS Number:  1405-87-4
Bacitracin

Bottle, 25 g

CAS Number:  1405-87-4
Price
531 zł
Log in for availability
1 * 25 g
Product Details & Documents

Bacitracin is active against gram-positive bacteria.

  • Approx. 75,000 units/g
  • CAS Number:1405-87-4
  • Formula:C₆₆H₁₀₃N₁₇O₁₆S₁
  • MDL Number:MFCD00062640
  • Molecular Weight:1421.60 g/mol
  • Melting Pt:221…225 °C
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.