Safety spectacles, Adaptec™
Anti-scratch, anti-fog, anti-static (narrow version), Black/red, Clear HS
numer katalogowy DALL1030739HS
Dostawca: HONEYWELL SAFETY
Przejdź do:
Product Details & Documents
Stylish single lens safety glasses with vented, cushioned frame and PC lens.
- Three position nose bridge is simple to adjust and keeps eyes centred in the lens
- Self-adjusting nose pads
- Adjustable lens angle reduces fogging; each temple hinge ratchets to seven positions
- Temple microadjustment allows workers to adjust temple tension without removing eyewear
- 4A+ coating on both sides of the lens ensures excellent anti-fog, anti-static and scratch resistant properties
Suitable for jobs where there are risks associated with abrasion, fogging, airborne particulates or impact.
Certifications: EN 166 FT
Dane techniczne
- Type:Anti-scratch, anti-fog, anti-static (narrow version)
- Colour:Black/red
- Seq:01
- Lenses:Clear HS