Order Entry
Poland
ContactUsLinkComponent
Heating Mantles WHM, for beakers 2000 ml, with integrated power controller, temperature range 450°C, 450 W, power supply 230 V , serial-no. 1 * 1 items
Catalog # WITGDH.WHM12147
Supplier:  witeg Labortechnik
Heating Mantles WHM, for beakers 2000 ml, with integrated power controller, temperature range 450°C, 450 W, power supply 230 V , serial-no. 1 * 1 items
Catalog # WITGDH.WHM12147
Supplier:  witeg Labortechnik
Frequently Bought Together